5-(4-Chlorophenyl)thiazol-2-amine 96%

Supplier: Ambeed
Warning

A228726-100MG A228726-250MG
103819-412EA 83 USD
103819-412 103850-316 76826-388 76826-390
5-(4-Chlorophenyl)thiazol-2-amine 96%
5-(4-Chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂S
MW: 210.68 g/mol
Boiling Pt: 389.8 °C (760 mm Hg)
Melting Pt: 203…204 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12779839
CAS Number: 73040-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR