5-(4-Bromophenyl)pyrimidine 97%

Supplier: Ambeed
Warning

A101072-100MG A101072-250MG A101072-1G
103795-790EA 248 USD
103795-790 77188-540 77188-542 77296-976
5-(4-Bromophenyl)pyrimidine 97%
5-(4-Bromophenyl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂
MW: 235.08 g/mol
Boiling Pt: 348.8±25.0 °C (760 mm Hg)
Flash Pt: 164.8 °C
Storage Temperature: Ambient
MDL Number: MFCD22422132
CAS Number: 160377-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR