5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Formyl-2-thiopheneboronic acid Pinacol ester, 2-(5-Formyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbaldehyde

A118648-10G A118648-1G A118648-5G
103799-086EA 13 USD
103799-086 76983-096 77311-754 77311-756
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde 98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅BO₃S
MW: 238.11 g/mol
Boiling Pt: 359.1±27.0 °C (760 mm Hg)
Flash Pt: 171 °C
Storage Temperature: Freezer
MDL Number: MFCD09266186
CAS Number: 1040281-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR