5-Pyrimidineboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(5-Pyrimidyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Pyrimidine-5-boronic acid pinacol ester, Pyrimidyl-5-boronic acid pinacol ester
A153887-100G
A153887-10G
A153887-1G
A153887-25G
A153887-5G
A153887-250MG
103804-576EA
10
USD
103804-576
103842-754
76689-470
76689-472
77300-034
77300-036
77318-438
77600-194
5-Pyrimidineboronic acid pinacol ester 97%
5-Pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₀H₁₅BN₂O₂ Boiling Pt: 308.9 °C (760 mm Hg) Melting Pt: 60…66 °C Flash Pt: 140.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05155223 CAS Number: 321724-19-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...