2-Amino-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Aminopyrimidine-5-boronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A117488-25G A117488-1G A117488-5G A117488-10G
103798-898EA 11 USD
103798-898 103824-786 77136-338 77311-568 77311-570 77614-920
2-Amino-5-pyrimidineboronic acid pinacol ester 98%
2-Amino-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₆BN₃O₂
MW: 221.06 g/mol
Boiling Pt: 394.8 °C (760 mm Hg)
Melting Pt: 209…211 °C
Flash Pt: 192.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR