5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-sulfonamide 97%

Supplier: Ambeed
Warning

A191891-1G A191891-100MG A191891-250MG
103816-408EA 570 USD
103816-408 77302-268 77683-502 77683-503
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-sulfonamide 97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-sulfonamide

Precaución: For research use only.

Formula: C₁₁H₁₇BN₂O₄S
MW: 284.14 g/mol
Boiling Pt: 470.0±55.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD13190590
CAS Number: 1083326-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR