2-Amino-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopyridine-3-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine, 2-Aminopyridine-5-boronic acid pinacol ester

A104258-100MG A104258-10G A104258-25G A104258-5G
103796-676EA 17 USD
103796-676 103822-386 77160-370 77160-372 77160-374 77309-254
2-Amino-5-pyridineboronic acid pinacol ester 97%
2-Amino-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₂
Boiling Pt: 352.9 °C (760 mm Hg)
Melting Pt: 138…141 °C
Flash Pt: 167.3 °C
Storage Temperature: Freezer
MDL Number: MFCD05663837
CAS Number: 827614-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR