5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyrimidin-2-amine 98%

Supplier: Ambeed
Warning

A347155-100MG A347155-250MG A347155-1G
103825-958EA 77 USD
103825-958 76817-274 76817-276 77306-580
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyrimidin-2-amine 98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₅BF₃N₃O₂
MW: 289.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09952051
CAS Number: 944401-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR