5-Indoleboronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Indole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
A198180-250MG
A198180-1G
A198180-5G
A198180-25G
103816-994EA
20
USD
103816-994
103847-620
76785-196
76785-198
77085-632
77085-634
77302-574
5-Indoleboronic acid pinacol ester 97%
5-Indoleboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₂ Boiling Pt: 396 °C (760 mm Hg) Melting Pt: 120…125 °C Flash Pt: 193.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD03789263 CAS Number: 269410-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...