1H-Indazole-5-boronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
A102724-10G
A102724-250MG
A102724-25G
A102724-5G
103796-420EA
118
USD
103796-420
103822-116
77008-406
77008-408
77008-410
77308-984
1H-Indazole-5-boronic acid pinacol ester 97%
1H-Indazole-5-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BN₂O₂ Melting Pt: 168…171 °C Flash Pt: 198.207 °C Storage Temperature: Freezer |
MDL Number:
MFCD07367523 CAS Number: 862723-42-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...