1H-Benzimidazole-5-boronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzimidazole, Benzimidazole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole
A181104-250MG
A181104-1G
A181104-100MG
103807-584EA
42
USD
103807-584
103845-950
76974-460
77083-468
77881-373
1H-Benzimidazole-5-boronic acid pinacol ester 95%
1H-Benzimidazole-5-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BN₂O₂ Melting Pt: 202…206 °C Storage Temperature: Freezer |
MDL Number:
MFCD11054041 CAS Number: 1007206-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...