5-(4'-(Bromomethyl)[1,1'-biphenyl]-2-yl)-1-trityl-1H-tetrazole 95%

Supplier: Ambeed
Danger

Synonyms: 5-(4'-(Bromomethyl)-[1,1'-biphenyl]-2-yl)-1-trityl-1H-tetrazole

A492130-5G A492130-100G A492130-1G A492130-500G A492130-25G
103839-298EA 6 USD
103839-298 76843-262 77143-958 77143-960 77143-962 77307-586
5-(4'-(Bromomethyl)[1,1'-biphenyl]-2-yl)-1-trityl-1H-tetrazole 95%
5-(4'-(Bromomethyl)[1,1'-biphenyl]-2-yl)-1-trityl-1H-tetrazole

Precaución: for Research Use Only

Formula: C₃₃H₂₅BrN₄
MW: 557.4824 g/mol
Boiling Pt: 718.4 °C (760 mm Hg)
Melting Pt: 152…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00665221
CAS Number: 124750-51-2
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR