5-(3-Bromophenyl)-1H-imidazole 95%

Supplier: Ambeed
Warning

A109053-100MG A109053-250MG A109053-5G
103797-436EA 175 USD
103797-436 76970-582 76970-584 76970-586
5-(3-Bromophenyl)-1H-imidazole 95%
5-(3-Bromophenyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Boiling Pt: 412.8 °C (760 mm Hg)
Flash Pt: 203.4 °C
Storage Temperature: Ambient
MDL Number: MFCD08688826
CAS Number: 53848-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR