5-(2-Hydroxyethyl)-3,4-dimethylthiazol-3-ium iodide 97%

Supplier: Ambeed
Warning

A206301-1G A206301-250MG A206301-25G A206301-5G
103817-704EA 9 USD
103817-704 103848-374 77086-622 77274-998 77275-000 77302-986
5-(2-Hydroxyethyl)-3,4-dimethylthiazol-3-ium iodide 97%
5-(2-Hydroxyethyl)-3,4-dimethylthiazol-3-ium iodide

Precaución: for Research Use Only

Formula: C₇H₁₂INOS
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00011958
CAS Number: 16311-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR