5-(2-Bromoethyl)-2,3-dihydrobenzofuran 98%

Supplier: Ambeed
Warning

A154248-10G A154248-1G A154248-25G A154248-5G
103804-618EA 16 USD
103804-618 103842-802 76689-482 77318-504 77318-506 77318-508
5-(2-Bromoethyl)-2,3-dihydrobenzofuran 98%
5-(2-Bromoethyl)-2,3-dihydrobenzofuran

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
Boiling Pt: 287.7 °C (760 mm Hg)
Melting Pt: 65…67 °C
Flash Pt: 113.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06797641
CAS Number: 127264-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR