5-(2,4-Dichlorophenyl)[1,3,4]thiadiazol-2-ylamine 95%

Supplier: Ambeed
Warning

Synonyms: 5-(2,4-Dichlorophenyl)-1,3,4-thiadiazol-2-amine

A139430-250MG A139430-1G A139430-5G
103802-738EA 108 USD
103802-738 77027-418 77299-076 77315-832
5-(2,4-Dichlorophenyl)[1,3,4]thiadiazol-2-ylamine 95%
5-(2,4-Dichlorophenyl)[1,3,4]thiadiazol-2-ylamine

Precaución: for Research Use Only

Formula: C₈H₅Cl₂N₃S
MW: 246.12 g/mol
Boiling Pt: 415.2 °C (760 mm Hg)
Flash Pt: 204.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00475836
CAS Number: 28004-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR