5-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-((tert-Butoxycarbonyl)amino)picolinic acid

A327716-250MG A327716-1G
103825-474EA 315 USD
103825-474 103836-424 76828-292 77306-360
5-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid 98%
5-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MW: 238.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09991768
CAS Number: 848308-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR