D-(+)-Biotin 98%

Supplier: Ambeed
Warning

Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7

A122167-100G A122167-10G A122167-25G A122167-50MG A122167-1G A122167-5G A122167-500G
103800-184EA 12 USD
103800-184 76810-540 76810-542 76810-544 76810-546 77312-322 77312-324 77670-726
D-(+)-Biotin 98%
D-(+)-Biotin

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₃S
MW: 244.31 g/mol
Boiling Pt: 573.6 °C (760 mm Hg)
Melting Pt: 231…233 °C
Flash Pt: 300.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00005541
CAS Number: 58-85-5
Merck Index: 13,01231

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR