4-(Trifluoromethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(4-(trifluoromethoxy)phenyl)borane, (4-(Trifluoromethoxy)phenyl)metaboric acid, 4-(Trifluoromethoxy)benzeneboronic acid, 4-Trifluoromethoxyphenylboronic acid

A270155-1G A270155-25G A270155-5G A270155-100G A270155-10G A270155-500G
103812-988EA 5 USD
103812-988 103852-894 77092-658 77092-660 77092-662 77140-942 77140-944 77685-769
4-(Trifluoromethoxy)phenylboronic acid 98%
4-(Trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
Boiling Pt: 256.6 °C (760 mm Hg)
Melting Pt: 123…127 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD01074648
CAS Number: 139301-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR