4-Tolylboronic acid 98%
Supplier: Ambeed
|
Warning
|
A128587-1000G
A128587-100G
A128587-10G
A128587-500G
A128587-25G
A128587-5G
76767-052EA
247
USD
76767-052
76767-054
76767-056
76767-058
76767-060
77313-496
77313-498
4-Tolylboronic acid 98%
4-Tolylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₉BO₂ Boiling Pt: 275.2 °C (760 mm Hg) Melting Pt: 256…263 °C Flash Pt: 61.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039138 CAS Number: 5720-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...