1-(4-Carboxyphenyl)-2-thiourea 97%

Supplier: Ambeed
Danger

Synonyms: 4-Thioureidobenzoic acid

A179817-100G A179817-250MG A179817-25G A179817-1G A179817-5G
103807-472EA 18 USD
103807-472 103845-824 76690-506 76690-508 77083-298 77301-642 77301-644
1-(4-Carboxyphenyl)-2-thiourea 97%
1-(4-Carboxyphenyl)-2-thiourea

Precaución: for Research Use Only

Formula: C₈H₈N₂O₂S
MW: 196.23 g/mol
Boiling Pt: 394.3 °C (760 mm Hg)
Flash Pt: 192.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041187
CAS Number: 7366-56-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR