4-Tetrahydrothiopyranone-1,1-dioxide 98%

Supplier: Ambeed
Warning

A120236-10G A120236-250MG
103799-904EA 18 USD
103799-904 76971-454 77670-706
4-Tetrahydrothiopyranone-1,1-dioxide 98%
4-Tetrahydrothiopyranone-1,1-dioxide

Precaución: for Research Use Only

Formula: C₅H₈O₃S
Boiling Pt: 385.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00460698
CAS Number: 17396-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR