4-Pyridylmethanol 99% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 4-Pyridylmethanol, 4-(Hydroxymethyl)pyridine, 4-Pyridinemethanol, Pyridine-4-methanol, 4-Pyridylcarbinol

A138065-100G A138065-10G A138065-500G A138065-1G A138065-25G A138065-5G
103802-550EA 5 USD
103802-550 77109-690 77109-692 77109-694 77315-552 77315-554 77315-556
4-Pyridylmethanol 99% (by HPLC)
4-Pyridylmethanol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 107…110 °C (760 mm Hg)
Melting Pt: 52…56 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006442
CAS Number: 586-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR