4-(4-Pyridyl)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yl)benzaldehyde

A159726-100G A159726-100MG A159726-10G A159726-250MG A159726-1G A159726-25G A159726-5G
103805-280EA 23 USD
103805-280 103843-506 76720-074 76720-076 76720-078 76720-080 77319-408 77319-410 77319-412
4-(4-Pyridyl)benzaldehyde 98%
4-(4-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
Melting Pt: 85…89 °C
Flash Pt: 165.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02684110
CAS Number: 99163-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR