4-(3-Pyridyl)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-3-yl)benzaldehyde

A305979-25G A305979-250MG A305979-1G A305979-5G
103814-450EA 25 USD
103814-450 103835-618 76838-322 77154-256 77305-926 77305-928
4-(3-Pyridyl)benzaldehyde 95%
4-(3-Pyridyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₉NO
Melting Pt: 52…57 °C
Storage Temperature: Ambient
MDL Number: MFCD01631910
CAS Number: 127406-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR