4-(1H-Pyrazol-1-yl)aniline 97%
Supplier: Ambeed
|
Warning
|
A259347-250MG
A259347-1G
A259347-25G
A259347-5G
103812-434EA
14
USD
103812-434
103852-314
77014-166
77091-862
77091-864
77091-866
4-(1H-Pyrazol-1-yl)aniline 97%
4-(1H-Pyrazol-1-yl)aniline
Precaución: for Research Use Only
Formula:
C₉H₉N₃ Boiling Pt: 312 °C (760 mm Hg) Melting Pt: 54 °C Flash Pt: 142.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00626298 CAS Number: 17635-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...