4-Propylcyclohexanone 98%

Supplier: Ambeed
Warning

A152461-100G A152461-10G A152461-25G
103842-564EA 6 USD
103842-564 76973-164 76973-166 77318-182
4-Propylcyclohexanone 98%
4-Propylcyclohexanone

Precaución: for Research Use Only

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 202.6 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00058690
CAS Number: 40649-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR