4-Propylbiphenyl 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Propyl-1,1'-biphenyl
A247685-100G
A247685-500G
A247685-25G
A247685-5G
103811-788EA
9
USD
103811-788
103851-624
77031-422
77031-424
77304-424
77304-426
4-Propylbiphenyl 98%
4-Propylbiphenyl
Precaución: for Research Use Only
Formula:
C₁₅H₁₆ MW: 196.29 g/mol Boiling Pt: 304.4 °C (760 mm Hg) Melting Pt: 15…17 °C Flash Pt: 141.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00102114 CAS Number: 10289-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...