4-Cyano-4'-propylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Propyl-[1,1'-biphenyl]-4-carbonitrile

A375392-1G A375392-25G A375392-5G A375392-250MG
103826-562EA 7 USD
103826-562 103837-586 76840-868 76840-870 76840-872 77033-336
4-Cyano-4'-propylbiphenyl 98%
4-Cyano-4'-propylbiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₅N
MW: 221.2970 g/mol
Boiling Pt: 367.5 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD00505987
CAS Number: 58743-76-3
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR