4-Propoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A134800-100G A134800-10G A134800-25G A134800-5G
77285-468EA 79.31 USD
77285-468 77285-470 77298-728 77298-730
4-Propoxybenzaldehyde 97%
4-Propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 129…130 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014134
CAS Number: 5736-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR