Piperidin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Piperidinol, 4-Hydroxypiperidine

A100690-100G A100690-10G A100690-25G A100690-500G A100690-1000G A100690-5G
103807-760EA 157 USD
103807-760 103821-272 77282-534 77282-536 77282-538 77282-540 77296-812 77296-814
Piperidin-4-ol 98%
Piperidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 222 °C (760 mm Hg)
Melting Pt: 86…90 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005999
CAS Number: 5382-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR