4-Phenylpyridine 98%
Supplier: Ambeed
|
Warning
|
A174628-250MG
A174628-1G
A174628-25G
A174628-5G
A174628-10G
103806-962EA
9
USD
103806-962
103845-262
76799-496
77082-566
77082-568
77082-570
77593-094
4-Phenylpyridine 98%
4-Phenylpyridine
Precaución: for Research Use Only
Formula:
C₁₁H₉N MW: 155.2 g/mol Boiling Pt: 274…275 °C (760 mm Hg) Melting Pt: 69…73 °C Flash Pt: 111.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006420 CAS Number: 939-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...