4-Phenylpiperidine 97%

Supplier: Ambeed
Warning

A129207-100G A129207-250MG A129207-1G A129207-25G A129207-10G
103801-244EA 9 USD
103801-244 76870-642 76870-644 77313-614 77313-616 77566-510
4-Phenylpiperidine 97%
4-Phenylpiperidine

Precaución: For research use only.

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 255…257 °C (760 mm Hg)
Melting Pt: 61…65 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006002
CAS Number: 771-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR