4-Phenylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Phenyl-1H-imidazole

A236573-1G A236573-25G A236573-100G A236573-10G A236573-5G
103811-108EA 14 USD
103811-108 103850-882 77089-932 77089-934 77288-602 77288-604 77304-060
4-Phenylimidazole 98%
4-Phenylimidazole

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.18 g/mol
Melting Pt: 128…131 °C
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00005197
CAS Number: 670-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR