4-Phenylcyclohexanone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane

A223732-100G A223732-10G A223732-500G A223732-25G A223732-5G
103809-578EA 514 USD
103809-578 103849-940 77288-256 77288-258 77288-260 77303-668 77303-670
4-Phenylcyclohexanone 97%
4-Phenylcyclohexanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Boiling Pt: 294 °C (760 mm Hg)
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR