3-[1,1'-Biphenyl]-4-ylacrylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylcinnamic acid

A586234-10G A586234-1G A586234-25G A586234-5G
103828-886EA 12 USD
103828-886 103840-034 76790-436 76844-168 76844-170 76844-172
3-[1,1'-Biphenyl]-4-ylacrylic acid 95%
3-[1,1'-Biphenyl]-4-ylacrylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₂
MW: 224.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014010
CAS Number: 13026-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR