4-Phenyl-1-butanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Phenylbutan-1-ol

A129424-100G A129424-10G A129424-500G A129424-5G A129424-25G
76688-566EA 31 USD
76688-566 76688-568 76688-570 77298-340 77313-670
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol

Precaución: for Research Use Only

Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 140
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR