4-Phenyl-2-pyrrolidone 95%

Supplier: Ambeed
Warning

A264048-1G A264048-25G A264048-5G A264048-10G
103812-650EA 8 USD
103812-650 103852-542 77092-190 77092-192 77092-194 77571-638
4-Phenyl-2-pyrrolidone 95%
4-Phenyl-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
Boiling Pt: 362.3 °C (760 mm Hg)
Flash Pt: 212.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01687226
CAS Number: 1198-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR