4-Phenoxyphenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxydiphenyl ether, Hydroquinone monophenyl ether

A174377-100G A174377-10G A174377-25G A174377-5G
103845-238EA 5 USD
103845-238 76872-544 76872-546 77082-540 77301-330
4-Phenoxyphenol 97%
4-Phenoxyphenol

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 80…84 °C
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002331
CAS Number: 831-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR