4-Phenoxybenzoic acid 98%

Supplier: Ambeed
Warning

A284279-100G A284279-10G A284279-500G A284279-25G A284279-5G A284279-1000G
103834-766EA 5 USD
103834-766 76723-360 76723-362 76723-364 76837-294 76837-296 77668-262
4-Phenoxybenzoic acid 98%
4-Phenoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
MW: 214.22 g/mol
Boiling Pt: 364.2 °C (760 mm Hg)
Melting Pt: 163…165 °C
Flash Pt: 141.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002539
CAS Number: 2215-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR