4-Phenoxybenzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A440614-1G A440614-250MG
103827-662EA 63 USD
103827-662 103838-728 76842-490 77886-167
4-Phenoxybenzene-1,2-diamine 98%
4-Phenoxybenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09038592
CAS Number: 13940-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR