[4-(2-Phenylethyl)phenyl]amine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Phenethylaniline hydrochloride

A367508-1G A367508-5G A367508-250MG
103826-424EA 18 USD
103826-424 103837-434 76840-670 76840-672 77003-484
[4-(2-Phenylethyl)phenyl]amine hydrochloride 97%
[4-(2-Phenylethyl)phenyl]amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClN
Storage Temperature: Ambient
MDL Number: MFCD06801189
CAS Number: 71845-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR