4-Pentyn-1-ol 95%

Supplier: Ambeed
Warning

Synonyms: 1-Pentyn-5-ol

A262888-1G A262888-25G A262888-5G A262888-100G A262888-10G A262888-500G
103812-582EA 5 USD
103812-582 103852-478 77092-106 77092-108 77092-110 77289-240 77289-242 77289-244
4-Pentyn-1-ol 95%
4-Pentyn-1-ol

Precaución: for Research Use Only

Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 154 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002974
CAS Number: 5390-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR