4-Pentylbenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amylbenzoic acid, 4-n-Pentylbenzoic acid, 4-Pentylbenzoic acid

A876426-100G A876426-25G A876426-5G
103841-302EA 6 USD
103841-302 76776-858 76845-570 77308-480
4-Pentylbenzoic acid 97%
4-Pentylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 304.6 °C (760 mm Hg)
Melting Pt: 85…90 °C (760 mm Hg)
Flash Pt: 147.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00002572
CAS Number: 26311-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR