4-Oxo-4H-pyran-2,6-dicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: Chelidonic acid, 2,6-Dicarboxy-4-oxo-4H-pyran

A131954-250MG A131954-5G A131954-1G
103801-662EA 15 USD
103801-662 77109-304 77298-524 77314-194
4-Oxo-4H-pyran-2,6-dicarboxylic acid 95%
4-Oxo-4H-pyran-2,6-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄O₆
MW: 184.103 g/mol
Melting Pt: 265 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006577
CAS Number: 99-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR