4-Oxo-1,4-dihydroquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dihydro-4-oxoquinoline-3-carboxylic acid

A219765-1G A219765-5G A219765-10G
103818-814EA 6 USD
103818-814 103849-694 77088-254 77088-256 77748-350
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid 97%
4-Oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Boiling Pt: 358.4 °C (760 mm Hg)
Melting Pt: 269…270 °C
Flash Pt: 170.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00498984
CAS Number: 13721-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR