Chelidamic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,4-Dihydro-4-oxo-2,6-pyridinedicarboxylic acid, 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid
A181222-10G
A181222-1G
A181222-25G
A181222-100G
A181222-500G
103807-590EA
6
USD
103807-590
103845-956
76813-574
77301-710
77301-712
77671-554
77671-556
Chelidamic acid 97%
Chelidamic acid
Precaución: for Research Use Only
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Boiling Pt: 428.3 °C (760 mm Hg) Melting Pt: 267 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066478 CAS Number: 138-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...