4-Oxazolemethanamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A190152-100MG A190152-250MG A190152-5G
103816-266EA 178 USD
103816-266 77164-246 77892-226 77892-227
4-Oxazolemethanamine hydrochloride ≥95%
4-Oxazolemethanamine hydrochloride

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₇ClN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD08437648
CAS Number: 1072806-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR