4-Nitrophthalonitrile 98%

Supplier: Ambeed
Warning

A117358-10G A117358-25G A117358-1000G A117358-500G A117358-100G
103824-752EA 6 USD
103824-752 77271-288 77271-290 77311-530 77549-352 77549-354 77592-292
4-Nitrophthalonitrile 98%
4-Nitrophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₃N₃O₂
Boiling Pt: 389.9 °C (760 mm Hg)
Melting Pt: 142…144 °C
Flash Pt: 189.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00040301
CAS Number: 31643-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR