(4-Nitrophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A113634-100G A113634-10G A113634-25G A113634-500G A113634-1000G
103808-106EA 132 USD
103808-106 103824-102 77025-898 77025-900 77025-902 77025-904 77310-916
(4-Nitrophenyl)acetic acid 97%
(4-Nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄(NO₂))CH₂COOH
Melting Pt: 150…155 °C
Flash Pt: 171.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00007383
CAS Number: 104-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR